Assay Details
Target Gene Details
Entrez Gene ID: | 4916 |
Gene Name: | neurotrophic receptor tyrosine kinase 3 |
Gene Aliases: |
GP145-TrkC, TRKC, gp145(trkC) |
Location: |
Chr.15:87859749-88256796 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Target Gene Details
Entrez Gene ID: | 283738 |
Gene Name: | NTRK3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:88252730-88271066 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NTRK3-AS1 | NR_038229.1 | |||
AK097486.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv952021 | Chr.15:88252070 - 88257269 on Build GRCh38 | Deletion | NTRK3-AS1 NTRK3 |
nsv827427 | Chr.15:88254355 - 88258505 on Build GRCh38 | Gain | NTRK3-AS1 NTRK3 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |