Assay Details
Target Gene Details
Entrez Gene ID: | 54880 |
Gene Name: | BCL6 corepressor |
Gene Aliases: |
ANOP2, MAA2, MCOPS2 |
Location: |
Chr.X:40051246-40177390 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BCOR | NM_001123383.1 | 15 | 5983 | NP_001116855.1 |
NM_001123384.1 | 14 | 5929 | NP_001116856.1 | |
NM_001123385.1 | 15 | 6014 | NP_001116857.1 | |
NM_017745.5 | 15 | 5912 | NP_060215.4 | |
XM_005272616.1 | 15 | 6146 | XP_005272673.1 | |
XM_005272618.3 | 15 | 5812 | XP_005272675.1 | |
XM_005272619.3 | 14 | 6418 | XP_005272676.1 | |
XM_005272620.3 | 14 | 6316 | XP_005272677.1 | |
XM_006724536.3 | 15 | 7248 | XP_006724599.1 | |
XM_011543929.2 | 15 | 5922 | XP_011542231.1 | |
XM_011543930.1 | 15 | 5935 | XP_011542232.1 | |
XM_011543931.2 | 15 | 6045 | XP_011542233.1 | |
XM_017029615.1 | 15 | 7146 | XP_016885104.1 | |
XM_017029616.1 | 15 | 6520 | XP_016885105.1 | |
AK000292.1 | 7 | 1789 | BAA91061.1 | |
AK074286.1 | 11 | 4920 | BAB85037.1 | |
AK095360.1 | 10 | 2623 | ||
AK225581.1 | 7 | 1742 | ||
BC009675.2 | 6 | 1540 | AAH09675.2 | |
BC063536.1 | 4 | 1096 | AAH63536.1 | |
BC114220.1 | 14 | 5929 | AAI14221.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |