Assay Details
Target Gene Details
Entrez Gene ID: | 219285 |
Gene Name: | sterile alpha motif domain containing 9 like |
Gene Aliases: |
C7orf6, DRIF2, UEF1 |
Location: |
Chr.7:93130054-93148401 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SAMD9L | NM_001303496.1 | 5 | 6575 | NP_001290425.1 |
NM_001303497.1 | 6 | 6404 | NP_001290426.1 | |
NM_001303498.1 | 5 | 6140 | NP_001290427.1 | |
NM_001303500.1 | 4 | 6008 | NP_001290429.1 | |
NM_152703.3 | 5 | 6780 | NP_689916.2 | |
XM_005250193.3 | 6 | 6212 | XP_005250250.1 | |
XM_006715890.1 | 2 | 5581 | XP_006715953.1 | |
XM_011515903.2 | 6 | 6465 | XP_011514205.1 | |
XM_017011822.1 | 4 | 6529 | XP_016867311.1 | |
XM_017011823.1 | 5 | 6285 | XP_016867312.1 | |
AB095926.1 | 5 | 6748 | ||
AI139687.1 | 1 | 390 | ||
AL834232.1 | 1 | 2288 | CAD38910.1 |