Assay Details
Target Gene Details
Entrez Gene ID: | 10850 |
Gene Name: | C-C motif chemokine ligand 27 |
Gene Aliases: |
ALP, CTACK, CTAK, ESKINE, ILC, PESKY, SCYA27 |
Location: |
Chr.9:34661883-34662692 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCL27 | NM_006664.3 | NP_006655.1 | ||
AB010445.1 | BAA87046.1 | |||
AF082393.1 | AAD41238.1 | |||
AJ243542.1 | CAB46983.1 | |||
BC148263.1 | ||||
BM719357.1 | ||||
CA433929.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3620335 | Chr.9:33871811 - 34738198 on Build GRCh38 | Gain | UBE2R2 LOC114224 DCTN3 CNTFR CCL19 FAM205A SNORD121B RPP25L ARID3C CNTFR-AS1 DNAI1 ENHO LOC730098 CCL21 FAM219A KIF24 DCAF12 SNORD121A IL11RA UBAP1 UBAP2 SIGMAR1 KIAA1161 CCL27 NUDT2 GALT C9orf24 |
nsv1161857 | Chr.9:34537279 - 34669174 on Build GRCh38 | Deletion | SIGMAR1 LOC730098 DCTN3 CCL27 GALT IL11RA RPP25L ARID3C CNTFR-AS1 CNTFR |
dgv12659n54 | Chr.9:34552439 - 34663141 on Build GRCh38 | Loss | SIGMAR1 DCTN3 CCL27 GALT IL11RA RPP25L ARID3C CNTFR-AS1 CNTFR |
nsv831544 | Chr.9:34650003 - 34818951 on Build GRCh38 | Gain | CCL19 PHF24 LOC730098 FAM205A CCL21 CCL27 GALT IL11RA |
nsv818699 | Chr.9:34535211 - 34685943 on Build GRCh38 | Gain | SIGMAR1 LOC730098 DCTN3 CCL27 GALT IL11RA RPP25L ARID3C CNTFR-AS1 CNTFR |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |