Assay Details
Target Gene Details
Entrez Gene ID: | 8848 |
Gene Name: | TSC22 domain family member 1 |
Gene Aliases: |
Ptg-2, TGFB1I4, TSC22 |
Location: |
Chr.13:44432143-44576565 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TSC22D1 | NM_001243797.1 | 3 | 672 | NP_001230726.1 |
NM_001243798.1 | 3 | 677 | NP_001230727.1 | |
NM_001243799.1 | NP_001230728.1 | |||
NM_006022.3 | 3 | 1014 | NP_006013.1 | |
NM_183422.3 | 3 | 3996 | NP_904358.2 | |
AB082525.1 | 3 | 3996 | ||
AJ222700.1 | 3 | 909 | CAA10951.1 | |
AK027071.1 | 3 | 2551 | ||
AK091854.1 | ||||
AK290751.1 | 3 | 949 | ||
BC000456.2 | 3 | 930 | AAH00456.1 | |
BC016867.1 | 3 | 933 | AAH16867.1 | |
BC069207.1 | 3 | 922 | AAH69207.1 | |
BC105277.1 | 3 | 925 | AAI05278.1 | |
BC136475.1 | 3 | 3766 | ||
BC136482.1 | 3 | 3766 | ||
BC146664.1 | 3 | 929 | ||
BC146679.1 | 3 | 929 | ||
BF307401.1 | 3 | 674 | ||
BI463370.1 | ||||
CR627459.1 | 4 | 2189 | CAH10539.1 | |
U35048.1 | 3 | 900 | AAC50566.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |