Assay Details
Target Gene Details
Entrez Gene ID: | 90639 |
Gene Name: | COX19, cytochrome c oxidase assembly factor |
Gene Aliases: |
- |
Location: |
Chr.7:964850-975599 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
COX19 | NM_001031617.2 | 3 | 4723 | NP_001026788.1 |
AA437231.1 | 1 | 174 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv605700 | Chr.7:886440 - 1065790 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv605699 | Chr.7:886312 - 1073232 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1028282 | Chr.7:964887 - 1000659 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv509182 | Chr.7:881218 - 1018010 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() ![]() |
nsv950784 | Chr.7:855964 - 1074664 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3571557 | Chr.7:823644 - 994725 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv605705 | Chr.7:947983 - 1211575 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv11111n54 | Chr.7:873118 - 983092 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
esv3571558 | Chr.7:950061 - 968142 on Build GRCh38 | Loss |
![]() ![]() |
nsv1161477 | Chr.7:928825 - 1077800 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)