Assay Details
Target Gene Details
Entrez Gene ID: | 4015 |
Gene Name: | lysyl oxidase |
Gene Aliases: |
- |
Location: |
Chr.5:122063195-122078501 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOX | NM_002317.6 | 1 | 683 | NP_002308.2 |
AF039291.1 | 1 | 416 | AAD02130.1 | |
AK297709.1 | 2 | 307 | ||
AK298781.1 | ||||
AK311354.1 | 1 | 441 | ||
AK312269.1 | 1 | 469 | ||
BC074820.2 | 1 | 189 | AAH74820.1 | |
BC074872.2 | 1 | 189 | AAH74872.1 | |
BC089436.1 | 1 | 455 | AAH89436.1 | |
DA461370.1 | ||||
M94054.1 | 1 | 244 | AAA59525.1 | |
S45875.1 | 1 | 167 | AAB23549.1 | |
S78694.1 | 1 | 397 | AAB21243.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |