Assay Details
Target Gene Details
Entrez Gene ID: | 253152 |
Gene Name: | epoxide hydrolase 4 |
Gene Aliases: |
ABHD7, EH4, EPHXRP |
Location: |
Chr.1:92029976-92063536 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPHX4 | NM_173567.4 | 1 | 217 | NP_775838.3 |
AK074822.1 | 1 | 177 | BAC11230.1 | |
BC041475.1 | 1 | 192 | AAH41475.1 | |
DB475492.1 | 1 | 217 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1002730 | Chr.1:91836984 - 92099672 on Build GRCh38 | Gain | BRDT SETSIP TGFBR3 BTBD8 EPHX4 |
nsv524845 | Chr.1:91750168 - 92133148 on Build GRCh38 | Gain | BRDT SETSIP TGFBR3 BTBD8 EPHX4 |
nsv830581 | Chr.1:91851650 - 92045618 on Build GRCh38 | Loss | BRDT TGFBR3 EPHX4 |
esv2750842 | Chr.1:91891224 - 92114118 on Build GRCh38 | Gain | BRDT SETSIP TGFBR3 BTBD8 EPHX4 |
esv2714684 | Chr.1:91095800 - 93049130 on Build GRCh38 | Deletion | KIAA1107 GFI1 FAM69A RPL5 RPAP2 SNORA66 BTBD8 CDC7 EVI5 BRDT HSP90B3P SETSIP SNORD21 HFM1 GLMN TGFBR3 C1orf146 EPHX4 |
nsv1006318 | Chr.1:91750851 - 92142767 on Build GRCh38 | Gain | BRDT SETSIP TGFBR3 BTBD8 EPHX4 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |