Assay Details
Target Gene Details
Entrez Gene ID: | 8509 |
Gene Name: | N-deacetylase and N-sulfotransferase 2 |
Gene Aliases: |
HSST2, NST2 |
Location: |
Chr.10:73801911-73811831 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDST2 | NM_003635.3 | 15 | 3776 | NP_003626.1 |
XM_005270255.3 | 15 | 3739 | XP_005270312.1 | |
XM_005270256.3 | 12 | 1981 | XP_005270313.1 | |
XM_011540310.2 | 14 | 4345 | XP_011538612.1 | |
XM_017016857.1 | 15 | 3805 | XP_016872346.1 | |
AB208870.1 | 8 | 4304 | BAD92107.1 | |
AK293527.1 | ||||
AK303648.1 | 16 | 3574 | ||
BC035711.1 | 15 | 3529 | AAH35711.1 | |
U36601.1 | 13 | 2999 | AAC27120.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv46n21 | Chr.10:73790158 - 73933937 on Build GRCh38 | Loss | C10orf55 CAMK2G NDST2 ZSWIM8-AS1 ZSWIM8 PLAU |
nsv551517 | Chr.10:73799109 - 73898823 on Build GRCh38 | Loss | CAMK2G NDST2 ZSWIM8-AS1 ZSWIM8 |
nsv1035634 | Chr.10:73724401 - 73907300 on Build GRCh38 | Gain | SEC24C BMS1P4 CHCHD1 FUT11 CAMK2G NDST2 ZSWIM8-AS1 GLUD1P3 ZSWIM8 |
nsv831912 | Chr.10:73649351 - 73840445 on Build GRCh38 | Gain | SEC24C BMS1P4 CHCHD1 FUT11 AGAP5 SYNPO2L CAMK2G NDST2 ZSWIM8-AS1 GLUD1P3 ZSWIM8 |
nsv825468 | Chr.10:73715689 - 73922563 on Build GRCh38 | Gain | SEC24C C10orf55 BMS1P4 CHCHD1 FUT11 CAMK2G NDST2 ZSWIM8-AS1 GLUD1P3 ZSWIM8 PLAU |
nsv551516 | Chr.10:73749515 - 73847410 on Build GRCh38 | Loss | SEC24C CHCHD1 FUT11 CAMK2G NDST2 ZSWIM8-AS1 ZSWIM8 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |