Assay Details
Target Gene Details
Entrez Gene ID: | 1582 |
Gene Name: | cytochrome P450 family 8 subfamily B member 1 |
Gene Aliases: |
CP8B, CYP12 |
Location: |
Chr.3:42872192-42876141 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CYP8B1 | NM_004391.2 | 1 | 394 | NP_004382.2 |
AF090318.1 | AAC63037.1 | |||
AK310784.1 | 1 | 151 | ||
AK315330.1 | 1 | 133 | ||
BC067434.1 | 1 | 102 | AAH67434.1 | |
BC067441.1 | 1 | 102 | AAH67441.1 | |
BC067442.1 | 1 | 102 | AAH67442.1 | |
BC067444.1 | 1 | 102 | AAH67444.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |