Assay Details
Target Gene Details
Entrez Gene ID: | 3831 |
Gene Name: | kinesin light chain 1 |
Gene Aliases: |
KLC, KNS2, KNS2A |
Location: |
Chr.14:103629188-103701551 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLC1 | NM_001130107.1 | 16 | 2282 | NP_001123579.1 |
NM_182923.3 | 15 | 2151 | NP_891553.2 | |
AL133587.1 | 2 | 925 | ||
AY180165.3 | AAO62550.1 | |||
AY180168.1 | 5 | 488 | AAO62553.1 | |
BM988840.1 | 1 | 249 | ||
L04733.1 | 15 | 2107 | AAA16576.1 |
Target Gene Details
Entrez Gene ID: | 7517 |
Gene Name: | X-ray repair cross complementing 3 |
Gene Aliases: |
CMM6 |
Location: |
Chr.14:103697611-103715486 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
XRCC3 | NM_001100118.1 | NP_001093588.1 | ||
NM_001100119.1 | NP_001093589.1 | |||
NM_005432.3 | NP_005423.1 | |||
XM_005268046.2 | XP_005268103.1 | |||
XM_011537138.2 | XP_011535440.1 | |||
AF035586.1 | AAC05368.1 | |||
AK022829.1 | ||||
AK023646.1 | ||||
AK124498.1 | ||||
AK126706.1 | ||||
BC001036.1 | AAH01036.1 | |||
BC011725.2 | AAH11725.1 | |||
BT007417.1 | AAP36085.1 | |||
BX161398.1 | CAD61884.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3635603 | Chr.14:103624778 - 103714856 on Build GRCh38 | Gain | XRCC3 LOC107984648 KLC1 |
esv2749175 | Chr.14:103700180 - 103705264 on Build GRCh38 | Deletion | XRCC3 KLC1 |
nsv566023 | Chr.14:103595309 - 103731914 on Build GRCh38 | Loss | ZFYVE21 XRCC3 LOC107984648 KLC1 |
nsv952260 | Chr.14:103697564 - 103723263 on Build GRCh38 | Deletion | ZFYVE21 XRCC3 LOC107984648 KLC1 |
nsv527425 | Chr.14:103695926 - 103731914 on Build GRCh38 | Loss | ZFYVE21 XRCC3 LOC107984648 KLC1 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | BRF1 LOC105370687 MIR4710 RD3L C14orf180 GPR132 NUDT14 LOC102723354 INF2 LOC107984648 AKT1 MIR6765 KIF26A LINC00638 C14orf79 TMEM179 MARK3 LINC00637 TDRD9 JAG2 ADSSL1 MIR203B EIF5 CDCA4 PPP1R13B PACS2 SIVA1 PLD4 C14orf2 XRCC3 KLC1 LOC101929634 LOC105378183 LOC388022 LOC105370691 MIR203A AHNAK2 CKB LOC105370708 ZFYVE21 BAG5 APOPT1 SNORA28 ZBTB42 CEP170B TRMT61A BTBD6 ASPG |
esv2760013 | Chr.14:103681188 - 103946476 on Build GRCh38 | Loss | RD3L ZFYVE21 C14orf2 LINC00637 TDRD9 XRCC3 LOC107984648 KLC1 PPP1R13B |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |