Assay Details
Target Gene Details
Entrez Gene ID: | 5875 |
Gene Name: | Rab geranylgeranyltransferase alpha subunit |
Gene Aliases: |
PTAR3 |
Location: |
Chr.14:24265538-24271627 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RABGGTA | NM_004581.5 | NP_004572.3 | ||
NM_182836.2 | NP_878256.1 | |||
AK291347.1 | ||||
AK292613.1 | ||||
AK303247.1 | ||||
AK308715.1 | ||||
AL547259.3 | ||||
AU125148.1 | ||||
BC003093.1 | AAH03093.1 | |||
CB995791.1 | ||||
CR541797.1 | CAG46596.1 | |||
DA379718.1 | ||||
DB116988.1 | ||||
Y08200.1 | CAA69382.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |