Assay Details
Target Gene Details
Entrez Gene ID: | 254531 |
Gene Name: | lysophosphatidylcholine acyltransferase 4 |
Gene Aliases: |
AGPAT7, AYTL3, LPAAT-eta, LPEAT2 |
Location: |
Chr.15:34358888-34367196 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LPCAT4 | NM_153613.2 | NP_705841.2 | ||
XM_006720454.2 | XP_006720517.1 | |||
XM_011521422.1 | XP_011519724.1 | |||
AF007155.1 | AAC19156.1 | |||
AF529233.1 | AAP97722.1 | |||
AF542964.1 | AAN33178.1 | |||
AK290273.1 | ||||
AL832237.1 | ||||
AY734233.1 | AAU34184.1 | |||
BC024892.2 | AAH24892.2 | |||
BC092463.1 | AAH92463.1 | |||
BC144236.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv517751 | Chr.15:34355367 - 34579095 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv471682 | Chr.15:34268250 - 34590429 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1052620 | Chr.15:32612402 - 34538731 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv33337 | Chr.15:31689863 - 34575097 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv509567 | Chr.15:34345031 - 34396055 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() |
nsv1485 | Chr.15:34354006 - 34601539 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() |
esv2760024 | Chr.15:34250820 - 34696046 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3569431 | Chr.15:33564570 - 35080307 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv469547 | Chr.15:34302356 - 34465606 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
dgv4456n54 | Chr.15:32604149 - 34557061 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)