Assay Details
Target Gene Details
Entrez Gene ID: | 129642 |
Gene Name: | membrane bound O-acyltransferase domain containing 2 |
Gene Aliases: |
LPAAT, LPCAT4, LPEAT, LPLAT 2, OACT2 |
Location: |
Chr.2:8852690-9003750 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MBOAT2 | NM_001321265.1 | 12 | 1799 | NP_001308194.1 |
NM_001321266.1 | 11 | 1721 | NP_001308195.1 | |
NM_001321267.1 | 14 | 2022 | NP_001308196.1 | |
NM_138799.3 | 13 | 1945 | NP_620154.2 | |
NR_135598.1 | 14 | 1997 | ||
NR_135599.1 | 13 | 2264 | ||
NR_135600.1 | 13 | 1851 | ||
NR_135601.1 | 11 | 1640 | ||
NR_135602.1 | 12 | 1812 | ||
NR_135603.1 | 14 | 2038 | ||
NR_135604.1 | 13 | 1961 | ||
NR_135605.1 | 12 | 1773 | ||
NR_135606.1 | 13 | 1864 | ||
AK027321.2 | 13 | 1905 | BAC85105.1 | |
AK074779.1 | 3 | 765 | BAC11204.1 | |
AK309818.1 | ||||
BC016005.1 | 10 | 1624 | AAH16005.1 | |
BC146871.1 | 13 | 1889 | ||
BC157827.1 | 13 | 1889 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833359 | Chr.2:8810194 - 8990672 on Build GRCh38 | Loss | KIDINS220 MBOAT2 |
nsv517367 | Chr.2:8853375 - 9046387 on Build GRCh38 | Gain | MBOAT2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |