Assay Details
Target Gene Details
Entrez Gene ID: | 4597 |
Gene Name: | mevalonate diphosphate decarboxylase |
Gene Aliases: |
FP17780, MDDase, MPD, POROK7 |
Location: |
Chr.16:88651935-88663153 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MVD | NM_002461.2 | NP_002452.1 | ||
XM_011523086.2 | XP_011521388.1 | |||
XM_011523087.2 | XP_011521389.1 | |||
XM_011523088.2 | XP_011521390.1 | |||
XM_011523089.2 | XP_011521391.1 | |||
AY203927.1 | AAP34450.1 | |||
BC000011.2 | AAH00011.1 | |||
BT006930.1 | AAP35576.1 | |||
U49260.1 | AAC50440.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471113 | Chr.16:88650665 - 88764428 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1926 | Chr.16:88626290 - 88660544 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() |
esv3639554 | Chr.16:88641322 - 88654848 on Build GRCh38 | Loss |
![]() ![]() |
nsv833325 | Chr.16:88553738 - 88698194 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv952070 | Chr.16:88257295 - 88780192 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv482951 | Chr.16:88633593 - 90228345 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)