Assay Details
Target Gene Details
Entrez Gene ID: | 23351 |
Gene Name: | KH and NYN domain containing |
Gene Aliases: |
KIAA0323 |
Location: |
Chr.14:24423186-24441342 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KHNYN | NM_001290256.1 | 3 | 580 | NP_001277185.1 |
NM_001290257.1 | 3 | 515 | NP_001277186.1 | |
NM_015299.2 | 3 | 643 | NP_056114.1 | |
XM_005267474.4 | 3 | 599 | XP_005267531.1 | |
XM_011536590.2 | 3 | 749 | XP_011534892.1 | |
XM_017021114.1 | 3 | 515 | XP_016876603.1 | |
AB002321.1 | 3 | 580 | ||
AK291680.1 | 3 | 496 | ||
BC042554.1 | 3 | 608 | AAH42554.1 | |
BX161465.1 | 3 | 616 | CAD61924.1 | |
DA674601.1 | 3 | 515 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |