Assay Details
Target Gene Details
Entrez Gene ID: | 116444 |
Gene Name: | glutamate ionotropic receptor NMDA type subunit 3B |
Gene Aliases: |
GluN3B, NR3B |
Location: |
Chr.19:1000419-1009814 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GRIN3B | NM_138690.2 | 3 | 1792 | NP_619635.1 |
XM_017026243.1 | 2 | 1038 | XP_016881732.1 | |
AY507106.1 | 3 | 1792 | AAS87020.1 | |
AY507107.1 | 3 | 1792 | AAS87021.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9662 | Chr.19:893394 - 1317740 on Build GRCh38 | Gain | TMEM259 ARHGAP45 LOC105372235 MIDN C19orf24 CBARP R3HDM4 EFNA2 CNN2 SBNO2 LOC102725180 RNU6-9 CIRBP-AS1 GRIN3B GPX4 KISS1R ARID3A CIRBP POLR2E ATP5D ABCA7 RNU6-2 WDR18 STK11 |
nsv1160555 | Chr.19:887275 - 1346084 on Build GRCh38 | Deletion | MED16 C19orf24 CBARP R3HDM4 EFNA2 CNN2 CIRBP-AS1 GRIN3B GPX4 POLR2E TMEM259 ARHGAP45 LOC105372235 MIDN SBNO2 LOC102725180 RNU6-9 KISS1R ARID3A CIRBP ATP5D ABCA7 RNU6-2 WDR18 STK11 |
nsv578188 | Chr.19:923744 - 1059005 on Build GRCh38 | Loss | TMEM259 GRIN3B LOC105372235 ARID3A ABCA7 RNU6-2 WDR18 CNN2 |
nsv469845 | Chr.19:951640 - 1144486 on Build GRCh38 | Loss | TMEM259 SBNO2 ARHGAP45 GRIN3B GPX4 LOC105372235 ARID3A POLR2E ABCA7 RNU6-2 WDR18 CNN2 |
esv2758743 | Chr.19:951641 - 1544934 on Build GRCh38 | Gain+Loss | APC2 PCSK4 NDUFS7 C19orf24 CBARP EFNA2 CNN2 CIRBP-AS1 GRIN3B ADAMTSL5 GPX4 RPS15 POLR2E PLK5 DAZAP1 TMEM259 ARHGAP45 LOC105372235 MUM1 MIDN SBNO2 GAMT LOC102725180 ARID3A CIRBP C19orf25 REEP6 ATP5D ABCA7 RNU6-2 WDR18 STK11 |
nsv953938 | Chr.19:355801 - 1104501 on Build GRCh38 | Deletion | MED16 LOC101928450 GZMM R3HDM4 MIR3187 PTBP1 PRTN3 CNN2 PALM GRIN3B GPX4 TPGS1 POLR2E CFD PLPPR3 RNF126 BSG TMEM259 PRSS57 SHC2 ARHGAP45 ODF3L2 LOC105372235 FGF22 FSTL3 C2CD4C CDC34 AZU1 MADCAM1 HCN2 POLRMT RNU6-9 KISS1R ARID3A THEG ELANE MISP MIR4745 ABCA7 RNU6-2 WDR18 |
nsv470101 | Chr.19:401714 - 1008505 on Build GRCh38 | Loss | MED16 LOC101928450 GZMM R3HDM4 MIR3187 PTBP1 PRTN3 PALM GRIN3B TPGS1 CFD PLPPR3 RNF126 BSG PRSS57 SHC2 ODF3L2 LOC105372235 FGF22 FSTL3 C2CD4C CDC34 AZU1 MADCAM1 HCN2 POLRMT RNU6-9 KISS1R ARID3A ELANE MISP MIR4745 WDR18 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |