Assay Details
Target Gene Details
Entrez Gene ID: | 84179 |
Gene Name: | major facilitator superfamily domain containing 7 |
Gene Aliases: |
LP2561 |
Location: |
Chr.4:676205-689468 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MFSD7 | NM_001294341.1 | NP_001281270.1 | ||
NM_001294342.1 | NP_001281271.1 | |||
NM_032219.3 | NP_115595.2 | |||
XM_005272297.1 | XP_005272354.1 | |||
XM_005272299.4 | XP_005272356.1 | |||
XM_006713918.1 | XP_006713981.1 | |||
XM_006713919.1 | XP_006713982.1 | |||
XM_011513568.1 | XP_011511870.1 | |||
XM_011513571.1 | XP_011511873.1 | |||
XM_011513572.1 | XP_011511874.1 | |||
XM_011513573.2 | XP_011511875.1 | |||
XM_011513574.1 | XP_011511876.1 | |||
XM_011513575.2 | XP_011511877.1 | |||
XM_017008700.1 | XP_016864189.1 | |||
AK025922.1 | BAB15283.1 | |||
AK292010.1 | ||||
AY203936.1 | AAP34459.1 | |||
AY358401.1 | AAQ88767.1 | |||
BC030246.1 | AAH30246.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv593206 | Chr.4:632173 - 741362 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
esv2759217 | Chr.4:609419 - 937566 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv829834 | Chr.4:671688 - 781736 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv950325 | Chr.4:620112 - 796912 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv593208 | Chr.4:674211 - 688444 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv508986 | Chr.4:527899 - 756994 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv10413 | Chr.4:673415 - 690512 on Build GRCh38 | Gain |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)