Assay Details
Target Gene Details
Entrez Gene ID: | 254956 |
Gene Name: | MORN repeat containing 5 |
Gene Aliases: |
C9orf113, C9orf18 |
Location: |
Chr.9:122159661-122200088 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MORN5 | NM_001286828.1 | NP_001273757.1 | ||
NM_198469.3 | NP_940871.2 | |||
XM_005251877.4 | XP_005251934.1 | |||
XM_005251878.3 | XP_005251935.1 | |||
XM_006717034.3 | XP_006717097.1 | |||
XM_011518457.2 | XP_011516759.1 | |||
XM_011518458.1 | XP_011516760.1 | |||
XM_017014578.1 | XP_016870067.1 | |||
AI026684.1 | ||||
AI560944.1 | ||||
AK128877.1 | BAC87653.1 | |||
AK302081.1 | ||||
BC133690.1 | ||||
HY011325.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv6702 | Chr.9:122151820 - 122184094 on Build GRCh38 | Insertion | MORN5 NDUFA8 |
nsv831715 | Chr.9:122066612 - 122226720 on Build GRCh38 | Gain | MIR4478 MORN5 NDUFA8 LHX6 TTLL11 |