Assay Details
Target Gene Details
Entrez Gene ID: | 23322 |
Gene Name: | RPGRIP1 like |
Gene Aliases: |
CORS3, FTM, JBTS7, MKS5, NPHP8, PPP1R134 |
Location: |
Chr.16:53599239-53703934 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RPGRIP1L | XM_005255867.1 | 20 | 3086 | XP_005255924.1 |
XM_005255868.1 | 22 | 3200 | XP_005255925.1 | |
XM_011522970.1 | 22 | 3200 | XP_011521272.1 | |
XM_011522971.2 | 22 | 3200 | XP_011521273.1 | |
XM_011522973.2 | 22 | 3200 | XP_011521275.1 | |
XM_017023094.1 | 22 | 3200 | XP_016878583.1 | |
XM_017023095.1 | 21 | 3098 | XP_016878584.1 | |
XM_017023096.1 | 22 | 3200 | XP_016878585.1 | |
XM_017023098.1 | 8 | 2163 | XP_016878587.1 | |
XM_017023099.1 | 8 | 1529 | XP_016878588.1 | |
AB023222.1 | 16 | 2515 | BAA76849.1 | |
AK026552.1 | 7 | 1147 | ||
AK304790.1 | 21 | 3250 | ||
AK307929.1 | 20 | 3106 | ||
AK308390.1 | 21 | 3198 | ||
BC136433.1 | 21 | 3193 | ||
CR749645.1 | 21 | 3207 | CAH18439.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |