Assay Details
Target Gene Details
Entrez Gene ID: | 64374 |
Gene Name: | SIL1 nucleotide exchange factor |
Gene Aliases: |
BAP, MSS, ULG5 |
Location: |
Chr.5:138946720-139198376 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SIL1 | NM_001037633.1 | 11 | 1683 | NP_001032722.1 |
NM_022464.4 | 10 | 1595 | NP_071909.1 | |
XM_011543570.2 | 12 | 1720 | XP_011541872.1 | |
AF547994.1 | 10 | 1514 | AAN84477.1 | |
AJ299442.1 | CAC17773.1 | |||
AK074624.1 | BAC11096.1 | |||
AK075177.1 | BAC11452.1 | |||
AY358950.1 | AAQ89309.1 | |||
BC011568.1 | AAH11568.1 | |||
CA415266.1 | 1 | 320 | ||
CD369479.1 | 1 | 320 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv950408 | Chr.5:138944112 - 138956011 on Build GRCh38 | Deletion | SIL1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |