Assay Details
Target Gene Details
Entrez Gene ID: | 55802 |
Gene Name: | decapping mRNA 1A |
Gene Aliases: |
HSA275986, Nbla00360, SMAD4IP1, SMIF |
Location: |
Chr.3:53283423-53347627 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DCP1A | NM_001290204.1 | 6 | 732 | NP_001277133.1 |
NM_001290205.1 | 8 | 901 | NP_001277134.1 | |
NM_001290206.1 | 6 | 718 | NP_001277135.1 | |
NM_001290207.1 | 5 | 604 | NP_001277136.1 | |
NM_018403.6 | 7 | 846 | NP_060873.4 | |
AB075507.1 | 7 | 762 | BAE45754.1 | |
AJ275986.1 | 7 | 829 | CAB77023.1 | |
AK001969.1 | 7 | 762 | BAA92008.1 | |
AK295205.1 | 6 | 630 | ||
AY146651.1 | 7 | 762 | AAN62763.1 | |
BC007439.2 | 7 | 737 | AAH07439.1 | |
BI769171.1 | ||||
KU178716.1 | 7 | 736 | ||
KU178717.1 | 6 | 622 | ||
KU178718.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3825 | Chr.3:53286923 - 53293922 on Build GRCh38 | Insertion | DCP1A |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |