Assay Details
Target Gene Details
Entrez Gene ID: | 57722 |
Gene Name: | immunoglobulin superfamily DCC subclass member 4 |
Gene Aliases: |
DDM36, NOPE |
Location: |
Chr.15:65381480-65423072 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IGDCC4 | NM_020962.2 | 20 | 5798 | NP_066013.1 |
XM_011521845.2 | 20 | 5978 | XP_011520147.1 | |
XM_011521846.2 | 20 | 5605 | XP_011520148.1 | |
XM_011521847.1 | 20 | 5665 | XP_011520149.1 | |
XM_017022448.1 | 20 | 5983 | XP_016877937.1 | |
AB046848.1 | 14 | 4858 | BAB13454.1 | |
AB052622.1 | 20 | 5798 | BAB86306.1 | |
AK124045.1 | 3 | 2721 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1036360 | Chr.15:65333881 - 65390204 on Build GRCh38 | Gain | IGDCC4 IGDCC3 |