Assay Details
Target Gene Details
Entrez Gene ID: | 4086 |
Gene Name: | SMAD family member 1 |
Gene Aliases: |
BSP-1, BSP1, JV4-1, JV41, MADH1, MADR1 |
Location: |
Chr.4:145481306-145559176 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SMAD1 | NM_001003688.1 | 2 | 304 | NP_001003688.1 |
NM_005900.2 | 2 | 480 | NP_005891.1 | |
XM_005262992.3 | 2 | 273 | XP_005263049.1 | |
XM_006714217.2 | 2 | 335 | XP_006714280.1 | |
XM_011531961.1 | 2 | 295 | XP_011530263.1 | |
XM_011531962.1 | 2 | 340 | XP_011530264.1 | |
XM_011531963.1 | 2 | 462 | XP_011530265.1 | |
XM_011531964.1 | 2 | 492 | XP_011530266.1 | |
AK293055.1 | 2 | 606 | ||
BC001878.1 | 2 | 405 | AAH01878.1 | |
BQ962660.1 | 2 | 372 | ||
BT007386.1 | 1 | 64 | AAP36050.1 | |
BU163870.1 | 2 | 491 | ||
U54826.1 | 2 | 304 | AAC50493.1 | |
U57456.1 | 1 | 216 | AAC50621.1 | |
U59423.1 | 2 | 496 | AAB06852.1 | |
U59912.1 | 2 | 305 | AAC50790.1 |
Target Gene Details
Entrez Gene ID: | 104326058 |
Gene Name: | SMAD1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.4:145514578-145517194 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SMAD1-AS1 | NR_126371.1 | 2 | 495 | |
AW087665.1 | 1 | 65 | ||
HY008085.1 | 2 | 496 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv1000240 | Chr.4:145514178 - 145523071 on Build GRCh38 | Deletion | SMAD1 SMAD1-AS1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |