Assay Details
Target Gene Details
Entrez Gene ID: | 1800 |
Gene Name: | dipeptidase 1 (renal) |
Gene Aliases: |
MBD1, MDP, RDP |
Location: |
Chr.16:89613308-89641540 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DPEP1 | NM_001128141.2 | NP_001121613.1 | ||
NM_004413.3 | NP_004404.1 | |||
XM_005256285.4 | XP_005256342.1 | |||
XM_005256286.3 | XP_005256343.1 | |||
XM_011522925.2 | XP_011521227.1 | |||
XM_017023009.1 | XP_016878498.1 | |||
AK309577.1 | ||||
BC017023.1 | AAH17023.1 | |||
BT006664.1 | AAP35310.1 | |||
CR457404.1 | CAG33685.1 | |||
D13138.1 | BAA02431.1 | |||
J05257.1 | AAB59410.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2762221 | Chr.16:89610380 - 90039318 on Build GRCh38 | Gain+Loss | SPATA33 LOC107984817 SPATA2L VPS9D1 CHMP1A AFG3L1P DBNDD1 VPS9D1-AS1 FANCA DPEP1 CENPBD1 CDK10 GAS8-AS1 ZNF276 TCF25 MC1R DEF8 GAS8 TUBB3 LOC105371419 SPIRE2 |
nsv952078 | Chr.16:89602793 - 89640192 on Build GRCh38 | Deletion | DPEP1 |
nsv1930 | Chr.16:89633876 - 89640750 on Build GRCh38 | Insertion | DPEP1 |
nsv428331 | Chr.16:89216635 - 90185980 on Build GRCh38 | Gain+Loss | ZNF778 SPATA2L VPS9D1 AFG3L1P FANCA FAM157C GAS8-AS1 ZNF276 MC1R DEF8 ANKRD11 LOC105371414 SPATA33 LOC107984817 TUBB8P7 LOC101927817 RPL13 URAHP CHMP1A SPG7 DBNDD1 VPS9D1-AS1 CPNE7 LOC100287036 DPEP1 CENPBD1 CDK10 TCF25 SNORD68 PRDM7 GAS8 TUBB3 LOC105371419 SPIRE2 |
nsv573739 | Chr.16:89625890 - 89696855 on Build GRCh38 | Gain | SPATA33 CDK10 SPATA2L CHMP1A DPEP1 |
nsv1063483 | Chr.16:89347250 - 89661187 on Build GRCh38 | Gain | SPATA33 LOC101927817 RPL13 CHMP1A SPG7 SNORD68 CPNE7 ANKRD11 DPEP1 |
nsv482951 | Chr.16:88633593 - 90228345 on Build GRCh38 | Loss | ZNF778 SPATA2L VPS9D1 AFG3L1P FANCA FAM157C LOC107987238 CDH15 GAS8-AS1 CBFA2T3 DEF8 LOC400558 LOC107984817 LOC101927817 URAHP CHMP1A SLC22A31 VPS9D1-AS1 PIEZO1 SNAI3-AS1 LOC101927793 SNORD68 PABPN1L TRAPPC2L TUBB3 GALNS LOC100129697 CYBA ACSF3 CDT1 RNF166 ZNF276 LOC339059 MC1R ANKRD11 LOC100289580 CTU2 LOC105371414 SPATA33 TUBB8P7 MIR4722 RPL13 APRT LINC00304 SPG7 DBNDD1 CPNE7 LOC100287036 SNAI3 LOC105371409 DPEP1 CENPBD1 IL17C CDK10 TCF25 PRDM7 MVD GAS8 LOC105371419 SPIRE2 |
nsv509639 | Chr.16:89513538 - 89775915 on Build GRCh38 | Insertion | SPATA33 RPL13 SPATA2L VPS9D1 CHMP1A SPG7 VPS9D1-AS1 CPNE7 FANCA DPEP1 CDK10 ZNF276 SNORD68 |
nsv1066382 | Chr.16:89537178 - 89661187 on Build GRCh38 | Gain | SPATA33 RPL13 CHMP1A SPG7 SNORD68 CPNE7 DPEP1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |