Assay Details
Target Gene Details
Entrez Gene ID: | 27230 |
Gene Name: | stress-associated endoplasmic reticulum protein 1 |
Gene Aliases: |
RAMP4 |
Location: |
Chr.3:150541993-150603177 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SERP1 | NM_014445.3 | NP_055260.1 | ||
XM_011512698.2 | XP_011511000.1 | |||
AB022427.1 | BAA81895.1 | |||
AF136975.1 | AAG49436.1 | |||
AK090933.1 | ||||
AK093929.1 | ||||
AK098135.1 | ||||
AK125413.1 | ||||
AL136807.1 | CAB66741.1 | |||
BC108314.1 | AAI08315.1 | |||
BC112364.1 | AAI12365.1 | |||
BC112365.1 | AAI12366.1 | |||
CR533472.1 | CAG38503.1 | |||
X75694.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs139300607] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Non-exonic non-DGV Variation |