Assay Details
Target Gene Details
Entrez Gene ID: | 3084 |
Gene Name: | neuregulin 1 |
Gene Aliases: |
ARIA, GGF, GGF2, HGL, HRG, HRG1, HRGA, MST131, MSTP131, NDF, NRG1-IT2, SMDF |
Location: |
Chr.8:31639222-32771716 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NRG1 | NM_001159995.2 | NP_001153467.1 | ||
NM_001159999.2 | NP_001153471.1 | |||
NM_001160001.2 | NP_001153473.1 | |||
NM_001322201.1 | NP_001309130.1 | |||
NM_001322202.1 | NP_001309131.1 | |||
NM_013962.2 | NP_039256.2 | |||
CN603652.1 | ||||
CN603654.1 | ||||
EF372273.1 | ||||
EF372274.1 | ||||
EF372275.1 | ||||
EF372276.1 | ||||
EF372277.1 | ||||
EF517295.1 | ||||
EF517296.1 | ||||
EF517297.1 | ||||
GQ983557.1 | ||||
GQ983558.1 | ||||
L12260.1 | AAB59622.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1076078 | Chr.8:31639683 - 31640284 on Build GRCh38 | Deletion | NRG1 |
esv3891380 | Chr.8:31278415 - 31706814 on Build GRCh38 | Gain | NRG1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |