Assay Details
Target Gene Details
Entrez Gene ID: | 9477 |
Gene Name: | mediator complex subunit 20 |
Gene Aliases: |
PRO0213, TRFP |
Location: |
Chr.6:41905353-41921147 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MED20 | NM_001305455.1 | 2 | 354 | NP_001292384.1 |
NM_001305456.1 | 4 | 626 | NP_001292385.1 | |
NM_001305457.1 | NP_001292386.1 | |||
NM_004275.4 | 3 | 509 | NP_004266.2 | |
NR_131160.1 | ||||
AF097725.1 | 3 | 461 | AAD16169.1 | |
AJ420544.1 | 2 | 312 | ||
AK023092.1 | 3 | 453 | BAB14399.1 | |
AK097385.1 | 4 | 626 | ||
AK293412.1 | ||||
AK300573.1 | 2 | 346 | ||
AK307070.1 | ||||
AL050196.1 | CAB43314.1 | |||
BC012618.1 | 3 | 505 | AAH12618.1 | |
BC019866.1 | 3 | 486 | ||
BC032552.1 | 3 | 471 | AAH32552.1 | |
BC040950.1 | 3 | 445 | AAH40950.1 | |
BU597139.1 | 3 | 504 | ||
DA909832.1 | 3 | 509 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |