Assay Details
Target Gene Details
Entrez Gene ID: | 283849 |
Gene Name: | exocyst complex component 3 like 1 |
Gene Aliases: |
EXOC3L |
Location: |
Chr.16:67184379-67190204 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EXOC3L1 | NM_178516.3 | NP_848611.2 | ||
AK092858.1 | BAC03991.1 | |||
AK092906.1 | BAC04000.1 | |||
AK292004.1 | ||||
AK300915.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain | CARMIL2 PSMB10 SLC12A4 E2F4 NUTF2 MIR328 FBXL8 C16orf86 LOC107984813 PSKH1 DPEP3 LRRC36 ELMO3 HSD11B2 SLC9A5 PLEKHG4 AGRP TRADD EXOC3L1 ENKD1 CTRL TPPP3 LOC100505942 RANBP10 TSNAXIP1 TMEM208 HSF4 KCTD19 ZDHHC1 KIAA0895L GFOD2 EDC4 THAP11 ACD CENPT DPEP2 FAM65A FHOD1 NOL3 ATP6V0D1 NRN1L PARD6A LOC105369155 DDX28 CTCF DUS2 LRRC29 LCAT LOC100131303 |
nsv827707 | Chr.16:67148355 - 67289296 on Build GRCh38 | Loss | C16orf70 FHOD1 ELMO3 NOL3 E2F4 TMEM208 HSF4 LOC105369155 SLC9A5 PLEKHG4 KIAA0895L B3GNT9 TRADD LRRC29 MIR328 EXOC3L1 FBXL8 |
nsv952044 | Chr.16:67141198 - 67244397 on Build GRCh38 | Deletion | C16orf70 FHOD1 ELMO3 NOL3 E2F4 TMEM208 HSF4 LOC105369155 KIAA0895L B3GNT9 TRADD LRRC29 MIR328 EXOC3L1 FBXL8 |
nsv1160429 | Chr.16:67161549 - 67220197 on Build GRCh38 | Deletion | HSF4 LOC105369155 KIAA0895L ELMO3 NOL3 LRRC29 MIR328 EXOC3L1 FBXL8 E2F4 |
nsv457513 | Chr.16:67181417 - 67282697 on Build GRCh38 | Loss | TMEM208 LOC105369155 SLC9A5 FHOD1 PLEKHG4 KIAA0895L ELMO3 LRRC29 MIR328 EXOC3L1 E2F4 |
nsv524492 | Chr.16:67174030 - 67294550 on Build GRCh38 | Loss | FHOD1 ELMO3 NOL3 E2F4 TMEM208 LOC105369155 KCTD19 SLC9A5 PLEKHG4 KIAA0895L LRRC29 MIR328 EXOC3L1 |
nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss | CARMIL2 PSMB10 SLC12A4 E2F4 NUTF2 B3GNT9 MIR328 FBXL8 C16orf86 LOC107984813 PSKH1 DPEP3 LRRC36 ELMO3 HSD11B2 SLC9A5 PLEKHG4 AGRP TRADD EXOC3L1 ENKD1 CTRL C16orf70 TPPP3 LOC100505942 RANBP10 TSNAXIP1 TMEM208 HSF4 KCTD19 ZDHHC1 KIAA0895L GFOD2 EDC4 THAP11 ACD CENPT DPEP2 FAM65A FHOD1 NOL3 ATP6V0D1 NRN1L PARD6A LOC105369155 DDX28 CTCF DUS2 LRRC29 LCAT LOC100131303 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |