Assay Details
Target Gene Details
Entrez Gene ID: | 6446 |
Gene Name: | serum/glucocorticoid regulated kinase 1 |
Gene Aliases: |
SGK |
Location: |
Chr.6:134169246-134318112 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SGK1 | NM_001143676.1 | NP_001137148.1 | ||
NM_001143677.1 | NP_001137149.1 | |||
NM_001143678.1 | NP_001137150.1 | |||
NM_001291995.1 | NP_001278924.1 | |||
NM_005627.3 | NP_005618.2 | |||
XM_011536071.1 | XP_011534373.1 | |||
AF153609.1 | AAD41091.1 | |||
AK055077.1 | ||||
AK098509.1 | ||||
AK293383.1 | ||||
AK295422.1 | ||||
AK298688.1 | ||||
BC001263.1 | AAH01263.1 | |||
BX649005.1 | 2 | 1461 | CAH10381.1 | |
DA309338.1 | ||||
DB450096.1 | ||||
EU518415.1 | ||||
FM205707.1 | ||||
FM205708.1 | ||||
FM205709.1 | ||||
FM205710.1 | ||||
Y10032.1 | CAA71138.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |