Assay Details
Target Gene Details
Entrez Gene ID: | 22933 |
Gene Name: | sirtuin 2 |
Gene Aliases: |
SIR2, SIR2L, SIR2L2 |
Location: |
Chr.19:38878555-38899862 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SIRT2 | NM_001193286.1 | NP_001180215.1 | ||
NM_012237.3 | NP_036369.2 | |||
NM_030593.2 | NP_085096.1 | |||
NR_034146.1 | ||||
XM_006723111.1 | XP_006723174.1 | |||
XM_011526654.1 | XP_011524956.1 | |||
XM_011526655.1 | XP_011524957.1 | |||
XM_017026500.1 | XP_016881989.1 | |||
AB009274.1 | ||||
AF083107.2 | AAD40850.2 | |||
AF095714.1 | AAD45971.1 | |||
AF131800.1 | AAD20046.1 | |||
AF160214.1 | AAF67015.1 | |||
AJ505014.1 | CAD43717.1 | |||
AK025876.1 | ||||
AK054642.1 | ||||
AK131390.1 | ||||
AK225824.1 | ||||
AY030277.1 | AAK51133.1 | |||
BC003012.1 | AAH03012.1 | |||
BC003547.1 | AAH03547.1 | |||
BF346568.1 | ||||
BF529906.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1160616 | Chr.19:38839331 - 38903590 on Build GRCh38 | Duplication | HNRNPL NFKBIB SIRT2 RINL |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |