Assay Details
Target Gene Details
Entrez Gene ID: | 688 |
Gene Name: | Kruppel like factor 5 |
Gene Aliases: |
BTEB2, CKLF, IKLF |
Location: |
Chr.13:73054976-73077542 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLF5 | NM_001286818.1 | NP_001273747.1 | ||
NM_001730.4 | 1 | 360 | NP_001721.2 | |
AB030824.1 | BAA96461.1 | |||
AF132818.1 | 1 | 360 | AAF18307.1 | |
AF287272.1 | 1 | 135 | AAF88068.1 | |
AK296900.1 | 1 | 150 | ||
AK302280.1 | ||||
AK304284.1 | 1 | 147 | ||
BC007695.1 | 1 | 138 | AAH07695.1 | |
BC042131.1 | AAH42131.1 | |||
EF208215.1 | ||||
HF546203.1 | 1 | 81 | ||
HF546204.1 | 1 | 81 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv22464 | Chr.13:73058544 - 73059901 on Build GRCh38 | Loss | KLF5 |
nsv820122 | Chr.13:73057729 - 73061960 on Build GRCh38 | Gain | KLF5 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |