Assay Details
Target Gene Details
Entrez Gene ID: | 4240 |
Gene Name: | milk fat globule-EGF factor 8 protein |
Gene Aliases: |
BA46, EDIL1, HMFG, HsT19888, MFG-E8, MFGM, OAcGD3S, SED1, SPAG10, hP47 |
Location: |
Chr.15:88898683-88913469 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MFGE8 | NM_001114614.2 | NP_001108086.1 | ||
NM_001310319.1 | NP_001297248.1 | |||
NM_001310320.1 | NP_001297249.1 | |||
NM_001310321.1 | NP_001297250.1 | |||
NM_005928.3 | NP_005919.2 | |||
XM_017022206.1 | XP_016877695.1 | |||
AK095908.1 | ||||
AK222735.1 | BAD96455.1 | |||
AK223182.1 | BAD96902.1 | |||
AK304627.1 | ||||
AK310357.1 | ||||
AK312640.1 | ||||
AU134066.1 | ||||
AY141974.1 | AAN08508.1 | |||
BC003610.2 | AAH03610.1 | |||
BT006948.1 | AAP35594.1 | |||
BX537974.1 | CAD97938.1 | |||
U58516.1 | AAC50549.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1653 | Chr.15:88870409 - 88915341 on Build GRCh38 | Deletion | HAPLN3 ACAN MFGE8 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |