Assay Details
Target Gene Details
Entrez Gene ID: | 49856 |
Gene Name: | WD repeat containing, antisense to TP73 |
Gene Aliases: |
WDR8 |
Location: |
Chr.1:3630767-3650107 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WRAP73 | NM_017818.3 | NP_060288.3 | ||
XM_005244754.1 | XP_005244811.1 | |||
XM_017001387.1 | XP_016856876.1 | |||
AB034912.1 | BAA92312.1 | |||
AK000437.1 | BAA91164.1 | |||
AK225525.1 | ||||
AK302397.1 | ||||
BC086311.1 | AAH86311.1 | |||
BX537591.1 | ||||
DC380547.1 | ||||
EF494669.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv410 | Chr.1:3635087 - 3667080 on Build GRCh38 | Insertion | TP73 WRAP73 |
nsv470687 | Chr.1:3356499 - 3763210 on Build GRCh38 | Loss | TP73-AS1 TPRG1L TP73 ARHGEF16 MIR551A CCDC27 MEGF6 WRAP73 PRDM16 |
nsv545128 | Chr.1:3630371 - 3700037 on Build GRCh38 | Loss | TP73 WRAP73 |
nsv1001715 | Chr.1:3627619 - 3769754 on Build GRCh38 | Loss | TP73-AS1 TPRG1L TP73 CCDC27 WRAP73 |
esv3585042 | Chr.1:3585575 - 4260083 on Build GRCh38 | Gain | TP73-AS1 MEGF6 WRAP73 DFFB LINC01134 TPRG1L TP73 CCDC27 SMIM1 LRRC47 LINC01346 CEP104 C1orf174 |
nsv1012423 | Chr.1:3477686 - 4747449 on Build GRCh38 | Gain | TP73-AS1 ARHGEF16 MEGF6 WRAP73 DFFB LINC01134 TPRG1L TP73 LOC284661 MIR551A CCDC27 SMIM1 LRRC47 LINC01346 CEP104 AJAP1 C1orf174 |
nsv951959 | Chr.1:3051037 - 3812036 on Build GRCh38 | Deletion | TP73-AS1 LOC105378604 ARHGEF16 MEGF6 WRAP73 LINC00982 MIR4251 PRDM16 TPRG1L TP73 MIR551A CCDC27 SMIM1 LRRC47 |
esv3585045 | Chr.1:3611161 - 3686920 on Build GRCh38 | Gain | TPRG1L TP73 MEGF6 WRAP73 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |