Assay Details
Target Gene Details
Entrez Gene ID: | 23033 |
Gene Name: | dopey family member 1 |
Gene Aliases: |
DOP1, KIAA1117, dJ202D23.2 |
Location: |
Chr.6:83067650-83171351 on Build GRCh38 |
Assay Gene Location: | Within Exon 41 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DOPEY1 | NM_001199942.1 | 40 | 7439 | NP_001186871.1 |
NM_015018.3 | 39 | 7406 | NP_055833.2 | |
XM_011535619.2 | 39 | 7431 | XP_011533921.1 | |
XM_017010559.1 | 40 | 7461 | XP_016866048.1 | |
XM_017010560.1 | 39 | 7389 | XP_016866049.1 | |
XM_017010561.1 | 40 | 7413 | XP_016866050.1 | |
XM_017010562.1 | 39 | 7362 | XP_016866051.1 | |
XM_017010563.1 | 39 | 7380 | XP_016866052.1 | |
XM_017010564.1 | 39 | 7380 | XP_016866053.1 | |
XM_017010565.1 | 39 | 7374 | XP_016866054.1 | |
XM_017010566.1 | 39 | 7374 | XP_016866055.1 | |
XM_017010567.1 | 38 | 7302 | XP_016866056.1 | |
XM_017010568.1 | 38 | 7371 | XP_016866057.1 | |
XM_017010569.1 | 37 | 7288 | XP_016866058.1 | |
XM_017010570.1 | 39 | 7362 | XP_016866059.1 | |
XM_017010571.1 | 39 | 7356 | XP_016866060.1 | |
XM_017010572.1 | 39 | 7311 | XP_016866061.1 | |
AB029040.2 | 22 | 4427 | BAA83069.2 | |
AK027030.1 | 19 | 3173 | BAB15631.1 | |
AK092929.1 | 8 | 1067 | ||
AL050080.1 | 11 | 1135 | CAB43259.1 | |
BC048342.1 | 13 | 1419 | AAH48342.1 | |
BC144544.1 | 39 | 7277 | ||
BC150619.1 | 39 | 7277 | ||
BX648667.1 | 39 | 7404 |
Target Gene Details
Entrez Gene ID: | 5238 |
Gene Name: | phosphoglucomutase 3 |
Gene Aliases: |
AGM1, IMD23, PAGM, PGM 3 |
Location: |
Chr.6:83150728-83193936 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PGM3 | NM_001199917.1 | 14 | 3201 | NP_001186846.1 |
NM_001199918.1 | 12 | 2863 | NP_001186847.1 | |
NM_001199919.1 | NP_001186848.1 | |||
NM_015599.2 | 13 | 3065 | NP_056414.1 | |
XM_017010935.1 | XP_016866424.1 | |||
XM_017010936.1 | XP_016866425.1 | |||
XM_017010937.1 | XP_016866426.1 | |||
XM_017010938.1 | XP_016866427.1 | |||
AK225363.1 | ||||
AL117443.1 | 4 | 1776 | CAB55928.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1026095 | Chr.6:83040907 - 83209453 on Build GRCh38 | Gain | DOPEY1 UBE3D PGM3 RWDD2A |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |