Assay Details
Target Gene Details
Entrez Gene ID: | 10495 |
Gene Name: | ecto-NOX disulfide-thiol exchanger 2 |
Gene Aliases: |
APK1, COVA1, tNOX |
Location: |
Chr.X:130622330-130903317 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ENOX2 | NM_001281736.1 | 14 | 3325 | NP_001268665.1 |
NM_006375.3 | 15 | 3469 | NP_006366.2 | |
NM_182314.2 | 16 | 3558 | NP_872114.1 | |
XM_005262354.3 | 15 | 3414 | XP_005262411.1 | |
XM_011531244.2 | 16 | 3560 | XP_011529546.1 | |
XM_011531245.2 | 16 | 3554 | XP_011529547.1 | |
XM_011531247.2 | 17 | 3686 | XP_011529549.1 | |
XM_011531249.2 | 14 | 3114 | XP_011529551.1 | |
XM_011531251.2 | 14 | 3219 | XP_011529553.1 | |
XM_017029226.1 | 18 | 3777 | XP_016884715.1 | |
XM_017029227.1 | 16 | 3511 | XP_016884716.1 | |
XM_017029228.1 | 15 | 3260 | XP_016884717.1 | |
XM_017029229.1 | 15 | 3463 | XP_016884718.1 | |
AF207881.2 | 13 | 3079 | AAF20934.2 | |
AK000353.1 | 14 | 3242 | ||
BC019254.1 | 10 | 2594 | AAH19254.2 | |
BC139781.1 | ||||
BC140874.1 | 14 | 3239 | ||
S72904.1 | 8 | 2235 | AAB30428.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73553287] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Exonic non-DGV Variation |