Assay Details
Target Gene Details
Entrez Gene ID: | 375607 |
Gene Name: | N-acetyltransferase 16 (putative) |
Gene Aliases: |
C7orf52 |
Location: |
Chr.7:101170493-101180276 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NAT16 | NM_198571.2 | 4 | 2896 | NP_940973.2 |
XM_005250325.1 | 5 | 2875 | XP_005250382.1 | |
XM_011516168.1 | 4 | 2857 | XP_011514470.1 | |
AK092119.1 | ||||
AK096556.1 | BAC04815.1 | |||
AL831911.1 | CAD38574.3 | |||
BU740913.1 | 1 | 58 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422376 | Chr.7:101100204 - 101373190 on Build GRCh38 | Duplication | VGF LOC101927721 AP1S1 IFT22 ZNHIT1 CLDN15 SERPINE1 LOC101927746 PLOD3 MOGAT3 FIS1 NAT16 MIR4653 COL26A1 |
nsv1030604 | Chr.7:101074451 - 101580185 on Build GRCh38 | Gain | VGF LOC101927721 AP1S1 IFT22 ZNHIT1 CLDN15 SERPINE1 LOC101927746 PLOD3 MOGAT3 LINC01007 TRIM56 FIS1 NAT16 MIR4653 COL26A1 |
dgv11534n54 | Chr.7:101160859 - 101238359 on Build GRCh38 | Loss | VGF PLOD3 AP1S1 MOGAT3 NAT16 ZNHIT1 CLDN15 |
esv2752134 | Chr.7:101156850 - 101498586 on Build GRCh38 | Gain | VGF LOC101927721 AP1S1 IFT22 ZNHIT1 CLDN15 LOC101927746 PLOD3 MOGAT3 FIS1 NAT16 MIR4653 COL26A1 |
nsv607973 | Chr.7:101139179 - 101230554 on Build GRCh38 | Loss | VGF SERPINE1 PLOD3 AP1S1 MOGAT3 NAT16 MIR4653 ZNHIT1 |
esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion | VGF EPHB4 TAF6 PILRA CLDN15 GPC2 PLOD3 MBLAC1 PMS2P1 GAL3ST4 AGFG2 CNPY4 C7orf43 ZCWPW1 TSC22D4 NAT16 MIR4653 AP1S1 ACHE MIR4658 AZGP1P1 GIGYF1 PPP1R35 MIR6875 NYAP1 ZNF3 SRRT POP7 AZGP1 ZAN LOC105375429 LRCH4 EPO PCOLCE-AS1 MEPCE STAG3 MIR106B FBXO24 ZNHIT1 ACTL6B C7orf61 SLC12A9 TFR2 ZASP TRIM56 MIR93 COPS6 LAMTOR4 GNB2 LOC102724094 MUC3A ZSCAN21 GATS AP4M1 MCM7 STAG3L5P-PVRIG2P-PILRB PILRB SAP25 MUC17 PVRIG MIR6840 MOSPD3 SERPINE1 ZKSCAN1 TRIP6 MOGAT3 PCOLCE UFSP1 FIS1 MUC12 PVRIG2P STAG3L5P SPDYE3 MIR25 |
nsv831075 | Chr.7:101049016 - 101273737 on Build GRCh38 | Loss | VGF SERPINE1 PLOD3 AP1S1 MOGAT3 TRIM56 FIS1 NAT16 MIR4653 MUC17 ZNHIT1 CLDN15 |
nsv607974 | Chr.7:101149529 - 101328323 on Build GRCh38 | Loss | VGF LOC101927746 LOC101927721 PLOD3 AP1S1 IFT22 MOGAT3 FIS1 NAT16 MIR4653 ZNHIT1 CLDN15 |
nsv1018841 | Chr.7:101117129 - 101225018 on Build GRCh38 | Gain | VGF SERPINE1 PLOD3 AP1S1 MOGAT3 NAT16 MIR4653 ZNHIT1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |