Assay Details
Target Gene Details
Entrez Gene ID: | 55830 |
Gene Name: | glycosyltransferase 8 domain containing 1 |
Gene Aliases: |
AD-017, MSTP139 |
Location: |
Chr.3:52694484-52706083 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GLT8D1 | NM_001010983.2 | 9 | 1652 | NP_001010983.1 |
NM_001278280.1 | 9 | 1648 | NP_001265209.1 | |
NM_001278281.1 | 9 | 1396 | NP_001265210.1 | |
NM_018446.3 | 8 | 1423 | NP_060916.1 | |
NM_152932.2 | 8 | 981 | NP_690909.1 | |
XM_017006857.1 | 10 | 1811 | XP_016862346.1 | |
XM_017006858.1 | 9 | 1582 | XP_016862347.1 | |
XM_017006859.1 | 8 | 1131 | XP_016862348.1 | |
AF157318.1 | 8 | 868 | AAF67484.1 | |
AF175227.1 | 9 | 1135 | AAQ13652.1 | |
AF176777.1 | 9 | 1138 | AAQ13655.1 | |
AJ271710.1 | 7 | 750 | CAC19666.1 | |
AK027517.1 | 8 | 889 | BAB55170.1 | |
AK075031.1 | 6 | 1285 | BAC11363.1 | |
AY358579.1 | 8 | 902 | AAQ88942.1 | |
BC119670.1 | 8 | 867 | ||
BC119671.1 | 8 | 867 | ||
CR749821.1 | 9 | 1105 | CAH18681.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |