Assay Details
Target Gene Details
Entrez Gene ID: | 56980 |
Gene Name: | PR domain 10 |
Gene Aliases: |
PFM7 |
Location: |
Chr.11:129899706-130003097 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRDM10 | NM_020228.2 | 18 | 2828 | NP_064613.2 |
NM_199437.1 | 17 | 2816 | NP_955469.1 | |
NM_199438.1 | 14 | 2554 | NP_955470.1 | |
NM_199439.1 | 13 | 2542 | NP_955471.1 | |
XM_011542912.2 | 17 | 3078 | XP_011541214.1 | |
XM_017018023.1 | 13 | 4627 | XP_016873512.1 | |
AB033057.2 | 14 | 2338 | BAA86545.2 | |
AF275817.1 | 14 | 2554 | AAF87243.1 | |
AF503171.1 | 13 | 2473 | AAP30847.1 | |
AK000234.1 | 11 | 1783 | BAA91026.1 | |
AK310354.1 | 19 | 2544 | ||
AL833172.1 | 17 | 2819 | ||
BC112934.1 | 17 | 2808 | AAI12935.1 | |
BC117415.1 | 14 | 2393 | ||
BC143612.1 | 14 | 2393 | ||
BC143613.1 | 13 | 2381 | ||
BX648944.1 | 18 | 2829 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv556564 | Chr.11:129908477 - 129934918 on Build GRCh38 | Gain | PRDM10 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |