Assay Details
Target Gene Details
Entrez Gene ID: | 346606 |
Gene Name: | monoacylglycerol O-acyltransferase 3 |
Gene Aliases: |
DC7, DGAT2L2, MGAT3 |
Location: |
Chr.7:101192886-101201131 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MOGAT3 | NM_001287147.1 | NP_001274076.1 | ||
NM_178176.3 | NP_835470.1 | |||
XM_005250309.3 | XP_005250366.1 | |||
XM_011516126.2 | XP_011514428.1 | |||
XM_011516127.2 | XP_011514429.1 | |||
AY229854.1 | AAO63579.1 | |||
AY358200.1 | AAQ88567.1 | |||
BC100953.1 | AAI00954.1 | |||
BC100954.1 | AAI00955.1 | |||
BC100955.1 | AAI00956.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422376 | Chr.7:101100204 - 101373190 on Build GRCh38 | Duplication | LOC101927721 MIR4653 CLDN15 COL26A1 ZNHIT1 NAT16 FIS1 MOGAT3 IFT22 LOC101927746 SERPINE1 AP1S1 PLOD3 VGF |
esv2422326 | Chr.7:101190168 - 101356850 on Build GRCh38 | Duplication | LOC101927721 IFT22 LOC101927746 CLDN15 ZNHIT1 FIS1 PLOD3 MOGAT3 |
nsv1030604 | Chr.7:101074451 - 101580185 on Build GRCh38 | Gain | LOC101927721 MIR4653 CLDN15 COL26A1 ZNHIT1 NAT16 FIS1 MOGAT3 IFT22 LOC101927746 SERPINE1 TRIM56 AP1S1 LINC01007 PLOD3 VGF |
dgv11534n54 | Chr.7:101160859 - 101238359 on Build GRCh38 | Loss | CLDN15 ZNHIT1 NAT16 AP1S1 PLOD3 VGF MOGAT3 |
esv2752134 | Chr.7:101156850 - 101498586 on Build GRCh38 | Gain | LOC101927721 MIR4653 CLDN15 COL26A1 ZNHIT1 NAT16 FIS1 MOGAT3 IFT22 LOC101927746 AP1S1 PLOD3 VGF |
nsv607973 | Chr.7:101139179 - 101230554 on Build GRCh38 | Loss | MIR4653 SERPINE1 ZNHIT1 NAT16 AP1S1 PLOD3 VGF MOGAT3 |
esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion | TSC22D4 PCOLCE-AS1 ZSCAN21 TAF6 MIR6875 FIS1 PVRIG EPO ACTL6B PILRA AGFG2 MEPCE AP1S1 PLOD3 C7orf43 ZCWPW1 TRIP6 MIR4653 ZKSCAN1 C7orf61 PPP1R35 CLDN15 MBLAC1 MIR6840 PILRB SAP25 MOGAT3 EPHB4 FBXO24 COPS6 ACHE MIR93 MIR106B PMS2P1 NYAP1 GATS ZNF3 MUC3A UFSP1 AZGP1P1 NAT16 LOC105375429 ZAN MIR4658 SLC12A9 PVRIG2P STAG3L5P-PVRIG2P-PILRB MOSPD3 PCOLCE GPC2 SERPINE1 LRCH4 GIGYF1 ZASP MIR25 VGF GAL3ST4 GNB2 STAG3L5P TFR2 STAG3 LOC102724094 CNPY4 ZNHIT1 MUC12 MUC17 AP4M1 POP7 MCM7 TRIM56 AZGP1 SRRT SPDYE3 LAMTOR4 |
nsv831075 | Chr.7:101049016 - 101273737 on Build GRCh38 | Loss | MIR4653 CLDN15 SERPINE1 ZNHIT1 TRIM56 NAT16 AP1S1 MUC17 FIS1 PLOD3 VGF MOGAT3 |
nsv607974 | Chr.7:101149529 - 101328323 on Build GRCh38 | Loss | LOC101927721 MIR4653 IFT22 LOC101927746 CLDN15 ZNHIT1 NAT16 AP1S1 FIS1 PLOD3 VGF MOGAT3 |
nsv1018841 | Chr.7:101117129 - 101225018 on Build GRCh38 | Gain | MIR4653 SERPINE1 ZNHIT1 NAT16 AP1S1 PLOD3 VGF MOGAT3 |
nsv831076 | Chr.7:101185566 - 101368393 on Build GRCh38 | Loss | LOC101927721 IFT22 LOC101927746 CLDN15 COL26A1 ZNHIT1 FIS1 PLOD3 MOGAT3 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |