Assay Details
Target Gene Details
Entrez Gene ID: | 51524 |
Gene Name: | transmembrane protein 138 |
Gene Aliases: |
HSPC196 |
Location: |
Chr.11:61362001-61376502 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM138 | NM_016464.4 | 2 | 500 | NP_057548.1 |
NR_028473.1 | ||||
XM_006718585.3 | 2 | 140 | XP_006718648.1 | |
XM_006718586.2 | 2 | 140 | XP_006718649.1 | |
XM_006718588.3 | XP_006718651.1 | |||
XM_011545098.2 | 2 | 140 | XP_011543400.1 | |
XM_011545099.1 | 2 | 140 | XP_011543401.1 | |
XM_011545100.1 | 2 | 140 | XP_011543402.1 | |
XM_017017917.1 | 2 | 140 | XP_016873406.1 | |
AF151030.1 | 2 | 117 | AAF36116.1 | |
AF151032.1 | 2 | 114 | AAF36118.1 | |
AK027724.1 | ||||
AK303219.1 | 2 | 115 | ||
AK307371.1 | ||||
AL832879.1 | 2 | 350 | CAI46174.1 | |
BC005201.1 | 2 | 115 | AAH05201.1 | |
BI907037.1 | 2 | 500 | ||
CB956757.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |