Assay Details
Target Gene Details
Entrez Gene ID: | 26277 |
Gene Name: | TERF1 interacting nuclear factor 2 |
Gene Aliases: |
DKCA3, TIN2 |
Location: |
Chr.14:24239641-24242674 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TINF2 | NM_001099274.1 | 9 | 1775 | NP_001092744.1 |
NM_012461.2 | 6 | 2102 | NP_036593.2 | |
XM_011536642.2 | 5 | 1997 | XP_011534944.1 | |
XM_017021216.1 | 5 | 1307 | XP_016876705.1 | |
XM_017021217.1 | 5 | 899 | XP_016876706.1 | |
AF195512.1 | 6 | 2022 | AAF18439.1 | |
AK023166.1 | 4 | 2493 | BAB14440.1 | |
AK225379.1 | 6 | 2046 | ||
AK294119.1 | 8 | 1601 | ||
BC005030.1 | 9 | 1556 | AAH05030.1 | |
BC019343.1 | 9 | 1733 | AAH19343.1 | |
BX161478.1 | 4 | 1830 | CAD61933.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |