Assay Details
Target Gene Details
Entrez Gene ID: | 1800 |
Gene Name: | dipeptidase 1 (renal) |
Gene Aliases: |
MBD1, MDP, RDP |
Location: |
Chr.16:89613308-89641540 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DPEP1 | NM_001128141.2 | NP_001121613.1 | ||
NM_004413.3 | NP_004404.1 | |||
XM_005256285.4 | XP_005256342.1 | |||
XM_005256286.3 | XP_005256343.1 | |||
XM_011522925.2 | XP_011521227.1 | |||
XM_017023009.1 | XP_016878498.1 | |||
AK309577.1 | ||||
BC017023.1 | AAH17023.1 | |||
BM152944.1 | ||||
J05257.1 | AAB59410.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2762221 | Chr.16:89610380 - 90039318 on Build GRCh38 | Gain+Loss | FANCA GAS8-AS1 VPS9D1-AS1 LOC105371419 TUBB3 SPATA33 VPS9D1 DBNDD1 TCF25 CHMP1A SPATA2L AFG3L1P SPIRE2 GAS8 DPEP1 DEF8 ZNF276 CDK10 MC1R LOC107984817 CENPBD1 |
nsv952078 | Chr.16:89602793 - 89640192 on Build GRCh38 | Deletion | DPEP1 |
nsv428331 | Chr.16:89216635 - 90185980 on Build GRCh38 | Gain+Loss | GAS8-AS1 VPS9D1-AS1 TUBB3 RPL13 SPATA33 SPATA2L DPEP1 DEF8 CPNE7 LOC101927817 ANKRD11 MC1R PRDM7 LOC107984817 SNORD68 FANCA LOC105371419 VPS9D1 LOC100287036 DBNDD1 TCF25 CHMP1A AFG3L1P ZNF778 SPIRE2 GAS8 TUBB8P7 SPG7 FAM157C LOC105371414 ZNF276 CDK10 URAHP CENPBD1 |
nsv573739 | Chr.16:89625890 - 89696855 on Build GRCh38 | Gain | SPATA33 CHMP1A SPATA2L CDK10 DPEP1 |
nsv1063483 | Chr.16:89347250 - 89661187 on Build GRCh38 | Gain | RPL13 SPATA33 LOC101927817 SPG7 CHMP1A ANKRD11 SNORD68 DPEP1 CPNE7 |
nsv482951 | Chr.16:88633593 - 90228345 on Build GRCh38 | Loss | ACSF3 LOC400558 CTU2 TRAPPC2L CYBA SPATA2L LINC00304 DPEP1 CPNE7 CDT1 RNF166 MIR4722 MC1R LOC107984817 SNORD68 FANCA SNAI3-AS1 LOC105371419 SLC22A31 VPS9D1 LOC100287036 TCF25 CHMP1A ZNF778 LOC105371409 MVD SPG7 LOC107987238 ZNF276 URAHP CENPBD1 GAS8-AS1 VPS9D1-AS1 TUBB3 RPL13 SPATA33 DEF8 CBFA2T3 LOC101927817 ANKRD11 SNAI3 PRDM7 LOC100289580 GALNS CDH15 DBNDD1 LOC339059 PABPN1L AFG3L1P LOC100129697 SPIRE2 GAS8 IL17C TUBB8P7 PIEZO1 LOC101927793 FAM157C LOC105371414 CDK10 APRT |
nsv509639 | Chr.16:89513538 - 89775915 on Build GRCh38 | Insertion | FANCA VPS9D1-AS1 RPL13 SPATA33 VPS9D1 CHMP1A SPATA2L DPEP1 CPNE7 SPG7 ZNF276 CDK10 SNORD68 |
nsv1066382 | Chr.16:89537178 - 89661187 on Build GRCh38 | Gain | RPL13 SPATA33 SPG7 CHMP1A SNORD68 DPEP1 CPNE7 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |