Assay Details
Target Gene Details
Entrez Gene ID: | 10344 |
Gene Name: | C-C motif chemokine ligand 26 |
Gene Aliases: |
IMAC, MIP-4a, MIP-4alpha, SCYA26, TSC-1 |
Location: |
Chr.7:75769524-75791583 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCL26 | NM_006072.4 | NP_006063.1 | ||
XM_017011671.1 | XP_016867160.1 | |||
XM_017011672.1 | XP_016867161.1 | |||
AA926779.1 | ||||
AB010447.1 | BAA84579.1 | |||
AB016542.1 | BAA36704.1 | |||
AF096296.1 | AAF09265.1 | |||
AF124601.1 | AAD22197.1 | |||
AF142434.1 | AAD46495.1 | |||
AY358893.1 | AAQ89252.1 | |||
BC069394.1 | AAH69394.1 | |||
BC093773.1 | AAH93773.1 | |||
BC101665.1 | AAI01666.1 | |||
BG211121.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3613737 | Chr.7:75769270 - 75786583 on Build GRCh38 | Gain | CCL26 |
dgv6468n100 | Chr.7:75701432 - 75777782 on Build GRCh38 | Gain | CCL26 HIP1 |
nsv528969 | Chr.7:75074053 - 76920728 on Build GRCh38 | Gain | SPDYE5 PMS2P2 YWHAG LOC100133091 STAG3L1 SPDYE16 ZP3 NCF1C PMS2P3 MDH2 LOC441259 PMS2P10 POM121C NSUN5P1 POR GTF2IP7 GTF2IP1 DTX2 STYXL1 HSPB1 TRIM73 LOC107986711 SRRM3 CCL26 POMZP3 SNORA14A SSC4D MIR4651 RHBDD2 FDPSP2 HIP1 TMEM120A CCL24 GTF2IRD2B LOC541473 LOC105375355 |
nsv518028 | Chr.7:75539242 - 75825134 on Build GRCh38 | Gain | CCL26 CCL24 HIP1 |
esv3613726 | Chr.7:75699055 - 75796413 on Build GRCh38 | Gain | CCL26 HIP1 |
esv2764051 | Chr.7:75663839 - 75875518 on Build GRCh38 | Gain+Loss | CCL26 CCL24 HIP1 |
dgv120n16 | Chr.7:75440769 - 76641919 on Build GRCh38 | Deletion | SPDYE5 YWHAG LOC100133091 SPDYE16 ZP3 PMS2P3 MDH2 POM121C POR GTF2IP7 DTX2 STYXL1 HSPB1 LOC107986711 SRRM3 CCL26 POMZP3 SNORA14A SSC4D MIR4651 RHBDD2 FDPSP2 HIP1 TMEM120A CCL24 LOC105375355 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |