Assay Details
Target Gene Details
Entrez Gene ID: | 256309 |
Gene Name: | coiled-coil domain containing 110 |
Gene Aliases: |
CT52, KM-HN-1, KMHN1 |
Location: |
Chr.4:185445182-185471759 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCDC110 | NM_001145411.1 | 5 | 650 | NP_001138883.1 |
NM_152775.3 | 6 | 761 | NP_689988.1 | |
XM_005262891.3 | 6 | 754 | XP_005262948.1 | |
XM_006714172.2 | 6 | 754 | XP_006714235.1 | |
XM_011531827.2 | 6 | 754 | XP_011530129.1 | |
XM_011531828.2 | 6 | 831 | XP_011530130.1 | |
XM_011531829.2 | 5 | 698 | XP_011530131.1 | |
XM_017007987.1 | 5 | 739 | XP_016863476.1 | |
XM_017007988.1 | 5 | 698 | XP_016863477.1 | |
AB080722.1 | 6 | 727 | BAC81776.1 | |
AK097605.1 | 6 | 754 | BAC05116.1 | |
AK302815.1 | 6 | 700 | ||
AL833337.1 | 6 | 706 | ||
BC028419.1 | 6 | 726 | AAH28419.1 | |
BC038515.1 | 5 | 590 | AAH38515.1 | |
BX641119.1 | 6 | 733 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv596373 | Chr.4:185451672 - 185605031 on Build GRCh38 | Gain | LOC105377590 PDLIM3 SORBS2 CCDC110 |
dgv5493n100 | Chr.4:185446588 - 185605031 on Build GRCh38 | Gain | LOC105377590 PDLIM3 SORBS2 C4orf47 CCDC110 |
nsv1033523 | Chr.4:185394656 - 185473803 on Build GRCh38 | Gain | LOC105377590 UFSP2 LRP2BP ANKRD37 C4orf47 CCDC110 |
nsv4638 | Chr.4:185430219 - 185464782 on Build GRCh38 | Insertion | C4orf47 CCDC110 |
esv3603496 | Chr.4:185447616 - 185603060 on Build GRCh38 | Gain | LOC105377590 PDLIM3 SORBS2 C4orf47 CCDC110 |
nsv517035 | Chr.4:185445161 - 185600889 on Build GRCh38 | Gain+Loss | LOC105377590 PDLIM3 SORBS2 C4orf47 CCDC110 |
nsv527026 | Chr.4:185404275 - 185632326 on Build GRCh38 | Gain | LOC105377590 UFSP2 PDLIM3 SORBS2 C4orf47 CCDC110 |
esv2759309 | Chr.4:185391971 - 185553542 on Build GRCh38 | Loss | LOC105377590 UFSP2 LRP2BP PDLIM3 ANKRD37 C4orf47 CCDC110 |
nsv428456 | Chr.4:185328666 - 185492417 on Build GRCh38 | Gain+Loss | LOC105377590 SNX25 UFSP2 LRP2BP ANKRD37 C4orf47 CCDC110 |
esv3603497 | Chr.4:185448235 - 185466446 on Build GRCh38 | Gain | C4orf47 CCDC110 |
nsv1029738 | Chr.4:185450051 - 185515559 on Build GRCh38 | Gain | LOC105377590 PDLIM3 CCDC110 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |