Assay Details
Target Gene Details
Entrez Gene ID: | 79872 |
Gene Name: | Cbl proto-oncogene like 1 |
Gene Aliases: |
HAKAI, RNF188 |
Location: |
Chr.7:107743497-107764997 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CBLL1 | NM_001284291.1 | 2 | 564 | NP_001271220.1 |
NM_024814.3 | 2 | 564 | NP_079090.2 | |
NR_024199.2 | 2 | 564 | ||
XM_011516580.2 | 2 | 764 | XP_011514882.1 | |
XM_017012643.1 | 2 | 365 | XP_016868132.1 | |
XM_017012644.1 | 2 | 758 | XP_016868133.1 | |
XM_017012645.1 | 2 | 616 | XP_016868134.1 | |
AK026762.1 | 2 | 120 | BAB15544.1 | |
AK293352.1 | ||||
AK307992.1 | 2 | 119 | ||
BC027460.2 | 1 | 89 | AAH27460.2 | |
BC130529.1 | 2 | 116 | ||
BC130531.1 | 2 | 116 | ||
BC144176.1 | 2 | 116 | ||
BP201841.1 | 2 | 116 | ||
BP280978.1 | 2 | 425 | ||
DB033469.1 | 2 | 119 | ||
DB174309.1 | 2 | 426 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |