Assay Details
Target Gene Details
Entrez Gene ID: | 80313 |
Gene Name: | leucine rich repeat containing 27 |
Gene Aliases: |
- |
Location: |
Chr.10:132330063-132381506 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LRRC27 | NM_001143757.1 | NP_001137229.1 | ||
NM_001309474.1 | NP_001296403.1 | |||
NM_030626.2 | NP_085129.1 | |||
XM_006717983.2 | XP_006718046.1 | |||
XM_006717984.2 | XP_006718047.1 | |||
XM_011540208.1 | XP_011538510.1 | |||
XM_011540209.1 | XP_011538511.1 | |||
XM_011540210.2 | XP_011538512.1 | |||
XM_017016690.1 | XP_016872179.1 | |||
XM_017016694.1 | XP_016872183.1 | |||
AB051461.1 | ||||
AK091472.1 | ||||
AK093250.1 | BAC04108.1 | |||
AK097418.1 | ||||
AK301422.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3624988 | Chr.10:132338306 - 132366249 on Build GRCh38 | Gain | LRRC27 |
nsv1051555 | Chr.10:132295188 - 132415461 on Build GRCh38 | Gain | PWWP2B LRRC27 STK32C |
esv3624986 | Chr.10:132322882 - 132551350 on Build GRCh38 | Gain | INPP5A LOC105378568 PWWP2B LRRC27 STK32C C10orf91 LOC107984282 |
esv2761642 | Chr.10:132048518 - 132365985 on Build GRCh38 | Gain | DPYSL4 LRRC27 STK32C JAKMIP3 |
nsv1054934 | Chr.10:130334322 - 133158679 on Build GRCh38 | Gain | INPP5A LINC01167 LOC105378568 MIR378C NKX6-2 PPP2R2D LINC01168 PWWP2B ADGRA1 LRRC27 C10orf91 JAKMIP3 LINC01164 TCERG1L TCERG1L-AS1 DPYSL4 STK32C ADGRA1-AS1 BNIP3 LOC107984282 CFAP46 LINC01166 |
nsv1052405 | Chr.10:132336481 - 132390781 on Build GRCh38 | Gain | LRRC27 |
nsv7627 | Chr.10:132350184 - 132375206 on Build GRCh38 | Insertion | LRRC27 |
dgv121n27 | Chr.10:132345217 - 132477022 on Build GRCh38 | Loss | LOC105378568 PWWP2B LRRC27 C10orf91 |
esv3577872 | Chr.10:132342397 - 132412909 on Build GRCh38 | Gain | PWWP2B LRRC27 |
esv2743659 | Chr.10:132355745 - 132356245 on Build GRCh38 | Deletion | LRRC27 |