Assay Details
Target Gene Details
Entrez Gene ID: | 283849 |
Gene Name: | exocyst complex component 3 like 1 |
Gene Aliases: |
EXOC3L |
Location: |
Chr.16:67184379-67190204 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EXOC3L1 | NM_178516.3 | NP_848611.2 | ||
AK092858.1 | BAC03991.1 | |||
AK092906.1 | BAC04000.1 | |||
AK292004.1 | ||||
AK300915.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain | SLC12A4 PSMB10 ENKD1 KCTD19 NUTF2 ACD TPPP3 LOC100131303 DUS2 AGRP DPEP2 LOC100505942 FBXL8 ZDHHC1 GFOD2 LRRC29 THAP11 EXOC3L1 KIAA0895L NRN1L TMEM208 MIR328 NOL3 E2F4 SLC9A5 LOC107984813 CARMIL2 PLEKHG4 TRADD CTRL ELMO3 FAM65A DPEP3 DDX28 LOC105369155 FHOD1 TSNAXIP1 LRRC36 CENPT PSKH1 ATP6V0D1 C16orf86 RANBP10 PARD6A HSD11B2 HSF4 EDC4 LCAT CTCF |
nsv827707 | Chr.16:67148355 - 67289296 on Build GRCh38 | Loss | LOC105369155 MIR328 FHOD1 NOL3 E2F4 FBXL8 SLC9A5 C16orf70 PLEKHG4 TRADD B3GNT9 ELMO3 LRRC29 EXOC3L1 KIAA0895L HSF4 TMEM208 |
nsv952044 | Chr.16:67141198 - 67244397 on Build GRCh38 | Deletion | LOC105369155 MIR328 FHOD1 NOL3 E2F4 FBXL8 C16orf70 TRADD B3GNT9 ELMO3 LRRC29 EXOC3L1 KIAA0895L HSF4 TMEM208 |
nsv1160429 | Chr.16:67161549 - 67220197 on Build GRCh38 | Deletion | LOC105369155 MIR328 NOL3 E2F4 FBXL8 ELMO3 LRRC29 EXOC3L1 KIAA0895L HSF4 |
nsv457513 | Chr.16:67181417 - 67282697 on Build GRCh38 | Loss | LOC105369155 MIR328 FHOD1 PLEKHG4 E2F4 ELMO3 LRRC29 SLC9A5 EXOC3L1 KIAA0895L TMEM208 |
nsv524492 | Chr.16:67174030 - 67294550 on Build GRCh38 | Loss | LOC105369155 MIR328 FHOD1 NOL3 E2F4 SLC9A5 KCTD19 PLEKHG4 ELMO3 LRRC29 EXOC3L1 KIAA0895L TMEM208 |
nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss | SLC12A4 PSMB10 ENKD1 KCTD19 NUTF2 ACD B3GNT9 TPPP3 LOC100131303 DUS2 AGRP DPEP2 LOC100505942 FBXL8 ZDHHC1 C16orf70 GFOD2 LRRC29 THAP11 EXOC3L1 KIAA0895L NRN1L TMEM208 MIR328 NOL3 E2F4 SLC9A5 LOC107984813 CARMIL2 PLEKHG4 TRADD CTRL ELMO3 FAM65A DPEP3 DDX28 LOC105369155 FHOD1 TSNAXIP1 LRRC36 CENPT PSKH1 ATP6V0D1 C16orf86 RANBP10 PARD6A HSD11B2 HSF4 EDC4 LCAT CTCF |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |