Assay Details
Target Gene Details
Entrez Gene ID: | 84134 |
Gene Name: | translocase of outer mitochondrial membrane 40 like |
Gene Aliases: |
TOMM40B |
Location: |
Chr.1:161225939-161230746 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TOMM40L | NM_001286373.1 | NP_001273302.1 | ||
NM_001286374.1 | NP_001273303.1 | |||
NM_032174.5 | NP_115550.2 | |||
XM_006711572.2 | XP_006711635.1 | |||
XM_011510057.2 | XP_011508359.1 | |||
XM_017002481.1 | XP_016857970.1 | |||
XM_017002482.1 | XP_016857971.1 | |||
AK022832.1 | BAB14265.1 | |||
AK027704.1 | BAB55309.1 | |||
AK297831.1 | ||||
AK297848.1 | ||||
AL523459.3 | ||||
BC014946.1 | AAH14946.1 | |||
BX537951.1 | CAD97918.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |