Assay Details
Target Gene Details
Entrez Gene ID: | 27158 |
Gene Name: | NADPH dependent diflavin oxidoreductase 1 |
Gene Aliases: |
NR1, bA350O14.9 |
Location: |
Chr.9:137205667-137219361 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDOR1 | NM_001144026.2 | 14 | 2259 | NP_001137498.1 |
NM_001144027.2 | 13 | 2108 | NP_001137499.1 | |
NM_001144028.2 | 14 | 2211 | NP_001137500.1 | |
NM_014434.3 | 14 | 2232 | NP_055249.1 | |
XM_011518545.1 | 13 | 2147 | XP_011516847.1 | |
XM_011518547.1 | 14 | 2049 | XP_011516849.1 | |
XM_011518548.2 | 13 | 3449 | XP_011516850.1 | |
XM_017014627.1 | 14 | 2210 | XP_016870116.1 | |
AF199509.1 | 14 | 2168 | AAF25205.1 | |
AK026089.1 | 14 | 2231 | ||
AK074403.1 | 13 | 2069 | ||
AK225995.1 | 14 | 2230 | ||
BC015735.1 | 14 | 2168 | AAH15735.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv13010n54 | Chr.9:137191539 - 137233413 on Build GRCh38 | Loss | TPRN NDOR1 RNF208 RNF224 SLC34A3 CYSRT1 TMEM203 |
nsv616012 | Chr.9:137197680 - 137223834 on Build GRCh38 | Loss | TPRN NDOR1 RNF208 TMEM203 |
nsv616007 | Chr.9:137079368 - 137240666 on Build GRCh38 | Loss | ANAPC2 UAP1L1 LOC101930307 MAN1B1 LRRC26 SSNA1 MIR3621 RNF208 RNF224 CYSRT1 TMEM203 TPRN NDOR1 TMEM210 MAN1B1-AS1 DPP7 GRIN1 SLC34A3 |
dgv13009n54 | Chr.9:137173890 - 137327859 on Build GRCh38 | Loss | ANAPC2 NRARP STPG3 SSNA1 C9orf173-AS1 RNF208 RNF224 CYSRT1 TOR4A TMEM203 TPRN NDOR1 TUBB4B FAM166A NELFB EXD3 SLC34A3 |
nsv524322 | Chr.9:136795692 - 137482216 on Build GRCh38 | Loss | MAN1B1 SSNA1 ABCA2 MIR3621 NPDC1 C8G TPRN NOXA1 GRIN1 FUT7 ANAPC2 LCN12 LOC101930307 MIR4292 LRRC26 RNF224 C9orf139 ENTPD2 MAMDC4 PNPLA7 TMEM210 PTGDS C9orf142 EXD3 MIR4479 FBXW5 UAP1L1 NRARP MIR7114 C9orf173-AS1 NSMF LCNL1 CYSRT1 TUBB4B CCDC183-AS1 NELFB MAN1B1-AS1 SLC34A3 RABL6 SAPCD2 TRAF2 PHPT1 STPG3 CLIC3 ENTPD8 PRR31 RNF208 TOR4A TMEM203 NDOR1 FAM166A CCDC183 EDF1 DPP7 C9orf172 |
nsv951207 | Chr.9:137136949 - 137379148 on Build GRCh38 | Deletion | ANAPC2 NRARP STPG3 LRRC26 SSNA1 MIR3621 C9orf173-AS1 RNF208 RNF224 CYSRT1 TOR4A TMEM203 TPRN NDOR1 TUBB4B FAM166A TMEM210 NELFB EXD3 GRIN1 SLC34A3 |
nsv1161918 | Chr.9:137118319 - 137438026 on Build GRCh38 | Duplication | ANAPC2 NRARP STPG3 LRRC26 ENTPD8 SSNA1 MIR3621 C9orf173-AS1 RNF208 RNF224 CYSRT1 TOR4A TMEM203 TPRN NDOR1 TUBB4B FAM166A TMEM210 NELFB EXD3 NOXA1 GRIN1 SLC34A3 |
esv2422220 | Chr.9:136584135 - 137350098 on Build GRCh38 | Deletion | HSPC324 TMEM141 LCN8 MAN1B1 SSNA1 ABCA2 MIR3621 NPDC1 C8G TPRN GRIN1 FUT7 ANAPC2 LCN12 LOC101930307 MIR4292 LRRC26 RNF224 C9orf139 SNORA17B ENTPD2 MAMDC4 TMEM210 PTGDS C9orf142 EXD3 MIR4479 FBXW5 UAP1L1 NRARP C9orf173-AS1 LOC100128593 LINC01451 LCNL1 CYSRT1 MIR6722 TUBB4B SNORA17A SNHG7 CCDC183-AS1 FAM69B NELFB MAN1B1-AS1 SLC34A3 RABL6 SAPCD2 TRAF2 LCN6 PHPT1 STPG3 CLIC3 EGFL7 AGPAT2 PRR31 RNF208 TOR4A TMEM203 LCN15 LCN10 NDOR1 MIR126 FAM166A CCDC183 EDF1 DPP7 C9orf172 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |